Accessories for VWR® VIS and UV-VIS Spectrophotometers
Catalog #: 10037-472
Supplier: VWR International
Accessories for VWR® VIS and UV-VIS Spectrophotometers
Catalog #: 10037-472
Supplier: VWR International
Specifications
- Description:Cuvette, quartz, 10 mm path length - 2 pcs
- Cat. no.:10037-472
Specifications
About this item
Customers Who Bought This Also Bought
Methanol ≥99.9%, OmniSolv®, gradient grade for HPLC, Supelco®
Supplier: MilliporeSigma
$635.20 to $2,541.50
Acetonitrile ≥99.9%, OmniSolv® LC-MS for LC-MS, Supelco®
Supplier: MilliporeSigma
$1,313.27 to $5,149.78
Taq DNA Polymerase with Standard Taq Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
$99.00
NEB®5-alpha Competent <i>E.coli</i> (High Efficiency), New England Biolabs
Supplier: New England Biolabs (NEB)
$300.00