WebSeal™ Standard Mats and Sealing Tapes, Non sterile
Catalog #: 10036-242
Supplier: Thermo Scientific
Specifications
- Color:Blue
- Description:WebSeal Mats, 96-Well, Flat Base, Round Well, 7 mm
- Material:Silicone
- Supplier No.:60180-M111
Specifications
About this item
These sealing mats are placed directly on the microplate by hand or with an applicator.
Manufactured from pure silicone with wider or small penetration area for ease of use. Mats are dry heat autoclavable, provide excellent chemical compatibility, and resist coring and tearing. Pre-slit mats ensure easy penetration and reduce vacuum problems. Non sterile.
Customers Who Bought This Also Bought