Certified WebSeal™ Plates, Plastic, Non-Coated, Nonsterile, Thermo Scientific
Catalog #: 10036-216
Supplier: Thermo Scientific
Specifications
- Volume:1000 µl
- Description:96-Well, Deep Well Microplates, Round Wells
- Height:41.6 mm
- Well Shape:U-Shaped
- Supplier No.:60180-P201
Specifications
About this item
Manufactured from ultra-low bleed, high-purity basis resin
Fully lot tested by GC-MS and LC/MS for organic extractables.
Customers Who Bought This Also Bought