Order Entry
ContactUsLinkComponent
Bio-Cert sterile 1000uL filtered pipette tip
BRAND® Filtered Pipette Tips for Pipettes using 1000 µl tips, BrandTech®
Catalog #: 10034-082
Supplier:  Brandtech
   Low Manufacturing Impact
Bio-Cert sterile 1000uL filtered pipette tip
BRAND® Filtered Pipette Tips for Pipettes using 1000 µl tips, BrandTech®
Catalog #: 10034-082
Supplier:  Brandtech
   Low Manufacturing Impact

Specifications

  • Volume:
    50 - 1000 µl
  • Environmentally Preferable:
    Low Manufacturing Impact
  • Packed:
    10 TipBoxes
  • Sterile:
    +
  • Supplier No.:
    732734

Specifications

About this item

BRAND filtered 1000 µl pipette tips are a 70 mm long tip for pipettes with a blue color code, that removes contaminating aerosols when pipetting volumes 1 ml or smaller.

All tips are produced under cleanroom conditions. New environmentally friendly refillable TipBox design is rigid and allows partial loading of multichannels. Hinged lid can be rotated 180 degrees to be a lift-off design. Environmentally friendly Tip Refills reload empty TipBoxes, and have 20% less waste. Sterile TipRefills have an included transfer aid to ensure sterility.

Graduations at 250, 500 and 1000 µl for visual verification of volume. Non-self-sealing filters allow easy recovery of sample if filter is wetted. PE filters are free from additives.

Certifications: CE-IVD.
Ordering information: All rack-packed tips are DNA-, RNase-, ATP-, and endotoxin-free.
Customers Who Bought This Also Bought