Order Entry
ContactUsLinkComponent
Mag-Bind Plant Dna Plus Kit
Mag-Bind® Plant DNA Plus 96 Kit, Omega Bio-tek
Catalog #: CA10035-802
Supplier:  Omega Bio-Tek
CAS Number:  
Mag-Bind® Plant DNA Plus 96 Kit, Omega Bio-tek
Catalog #: CA10035-802
Supplier:  Omega Bio-Tek
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Magnetic beads
  • Prep size:
    Scalable
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
  • Target molecule:
    DNA
  • Description:
    Mag-Bind® Plant DNA Plus 96 Kit
  • Size:
    4 x 96 Preps
  • Description:
    Mag-Bind® Plant DNA Plus 96 Kit
  • Cat. no.:
    CA10035-802
  • Includes:
    Mag-Bind® Particles CNR, CSPL Buffer, CSPW1 Buffer, CSPW2 Buffer, SPM Wash Buffer, Elution Buffer, RNase A and User Manual
  • Supplier No.:
    M1128-01

Specifications

About this item

The Mag-Bind® Plant DNA 96 Kit allows rapid and reliable isolation of high-quality genomic DNA from a wide variety of difficult to lyse and high polysaccharide plant species and tissues.

  • Simple, rapid, and reliable procedure
  • Reproducible yields of pure, ready to use DNA
  • Adaptable to most robotic liquid handling platform

The system utilizes a CTAB based lysis buffer without the need for a chloroform or other organic solvent steps

The Mag-Bind® Plant DNA Kit also provides greater flexibility than silica plate based systems as partial plates can be used and can be scaled up to a 24-well format with the appropriate magnetic stand. Purified DNA is suitable for PCR, qPCR, restriction digestion, hybridization applications, and more demanding applications such as RAPD.

The propietary wash buffer systems eliminate polysaccharides, phenolic compounds, and enzyme inhibitors from plant tissue lysates. This kit is designed for manual or fully automated high throughput preparation of genomic, chloroplast, and mitochondrial DNA.

Customers Who Bought This Also Bought