Order Entry
ContactUsLinkComponent
4 X 6 ZIPCLEAN POUCHES
Precision Zip-Clean Pouches™, PPC Flexible Packaging
Catalog #: 10033-162
Supplier:  Fisher Container
Precision Zip-Clean Pouches™, PPC Flexible Packaging
Catalog #: 10033-162
Supplier:  Fisher Container

Specifications

  • Dimensions:
    10.2×15.2 cm (4×6")
  • Cat. no.:
    10033-162
  • Supplier No.:
    91085P

Specifications

About this item

Tough, ultraclean, high clarity film that is ideal for all multi-use, reclosable cleanroom and laboratory applications.

  • Easy open and easy zip closure
  • Cleanroom certified
  • Barrier properties

Sealed in a double-lined carton.

Customers Who Bought This Also Bought