Order Entry
ContactUsLinkComponent
FASTDNA SPIN KIT
FastDNA™ Spin Kit, MP Biomedicals
Catalog #: IC116540600
Supplier:  MP Biomedicals
CAS Number:  
FASTDNA SPIN KIT
FastDNA™ Spin Kit, MP Biomedicals
Catalog #: IC116540600
Supplier:  MP Biomedicals
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Method / Format:
    Column (silica-based)
  • Prep size:
    Mini
  • Refine target molecule:
    Total DNA
  • Sample type:
    Plant samples
    Yeast/Fungi
    Cultured cells
    Invertebrates
    Human/animal cells and tissues
  • Target molecule:
    DNA
  • Description:
    FastDNA™ Spin Kit
  • Size:
    100×2 ml preps
  • Description:
    FastDNA™ Spin Kit
  • Cat. no.:
    IC116540600
  • Supplier No.:
    116540600

Specifications

About this item

The Fast DNA® SPIN Kit is used with the FastPrep®-24 or FastPrep® FP120 instrument to lyse and subsequently isolate DNA from up to 200 mg of almost any sample in less than 30 minutes.

  • Isolation of genomic DNA from plants, animals, bacteria, yeast, algae, and fungi using a silica spin filter method
  • Rapid and reproducible sample lysis with the FastPrep®-24 or FastPrep® FP120 instrument
  • Lyse and isolate DNA in less than 30 minutes from a variety of sample types
  • Flexible format eliminates need for separate DNA isolation kits
  • No hazardous organic reagents required
  • SPIN filters are included to streamline silica handling

Lysing Matrix A and three different chaotropic buffers are provided for the optimal homogenisation of a wide variety of sample types (plant and animal tissues, cultured cells, bacteria, yeast, fungi, insects, etc). The released DNA is purified by a silica-based GeneClean® SPIN procedure, with supplied filter buckets and catch tubes to simplify liquid handling, and is suitable for PCR analysis and other downstream applications.

Instrument Compatibility: FastPrep-24, FastPrep 120, FastPrep-96, SuperFastPrep-1

Ordering information: Kit contains Lysing Matrix A, 100×2,0 ml tubes;1/4" Ceramic Spheres, 100 spheres; Binding Matrix, 66 ml; Concentrated SEWS-M, 12 ml; DES, 25 ml; CLS-VF, 90 ml; PPS, 25 ml; CLS-TC, 110 ml; CLS-Y, 110 ml; SPIN Modules, 100 each; and Catch Tubes, 100 each.
Materials required not supplied include FastPrep® Instrument or alternative high speed disruptor; Microcentrifuge that can freely spin 2.0 mL tubes; Microcentrifuge tubes (2.0 mL and 1.5 mL); Rotator or low-speed vortex.
Caution: For research purposes only.
Customers Who Bought This Also Bought