Screw Caps for Sample Tubes, with O-Ring, Globe Scientific
Catalog #: 10028-126
Supplier: Globe Scientific
Specifications
- Color:Gray
- Cat. no.:10028-126
- Supplier No.:6137A
Specifications
About this item
All caps include an O-Ring to ensure a leak-tight seal at most temperatures.
Customers Who Bought This Also Bought