n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC, VWR Chemicals BDH®
Catalog #: CABDH24575.400
Supplier: VWR International
CAS Number: 110-54-3
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber
- Size:4 L
- Cat. no.:CABDH24575.400
- Supplier No.:VWRCBDH24575.4
Specifications
Specification Test Results
- Assay (GC):Min. 97 %
- Water:Max. 0.005 %
- Non-volatile residue:Max. 0.0005 %
- Acidity:Max. 0.0003 meq/g
- Alkalinity:Max. 0.0002 meq/g
- Transmittance (210 nm):Min. 50 %
- Transmittance (220 nm):Min. 80 %
- Transmittance (245 nm):Min. 98 %
- Conforms to BDH 15249:Passes test
- VWR Chemicals BDH®, n-Hexane ≥97%, HiPerSolv CHROMANORM® for HPLC:
Specification Test Results
About this item
Synonyms: Hexyl hydride,Hexanen
:n-Hexane
:Storage Temperature: Ambient
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94.3 °C
Density:0.659 g/cm³ (20 °C)
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:97
:Filtered through a 0.2 µm filter, packaged under nitrogen.
Flash Pt:–22 °C
:Boiling Pt:
:Boiling Pt: 69 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –94.3 °C
:Flash Pt:
:Flash Pt: –22 °C
:M.W. 86.18 g/mol
Merck Index:13,04712
Storage Temperature:Ambient
EINECS:203-777-6
:69
UN:1208
:3
:II
:–22 °C
:GHS02
:GHS08
:GHS07
:GHS09
:Filter 0,2 um_Nitrogen_filled
Customers Who Bought This Also Bought