Specifications
- Capacity:100 ml
- Length:5.97 cm (2.35")
- Sterility:Pre-sterile
- Width:3.14 cm (1.24")
- Description:Disposable Reagent Reservoirs
- Material:Polystyrene
- Packaging:5/Bag, 10 Bags/Pack, 4 Packs per Case
- Supplier No.:SR-0100-5SNM
Specifications
About this item
Biotix disposable reagent reservoirs are designed for use in multi-channel pipetting applications and can accommodate up to 12-channel pipettors. Angled sides and a V-shaped bottom allow for maximum recovery of reagents. All reservoirs feature internal graduated volume lines and pour-spouts on the corners. Choose from opaque white polyvinyl chloride (PVC) or high clarity polystyrene (PS) in several packaging configurations.
Biotix disposable reagent reservoirs are designed for use in multi-channel pipetting applications and can accommodate up to 12-channel pipettors
Biotix Reagent reservoirs are certified free of DNA, DNase, RNase, and endotoxins (pyrogens). Manufactured under stringent controls in an ISO 9001 certified facility. Every lot is tested for engineering compliance and accuracy. All raw materials are traceable.
How Biotix Reservoirs can help in the lab:
Designed for use in multi-channel pipetting applications and fully compatible with 12-channel pipettors
Angled sides and a V-shaped bottom allow for maximum reagent recovery
Manufactured with internal volume graduation lines and pour-spouts on the corners
Available in polyvinyl chloride (PVC) or clear polystyrene (PS)
Angled sides and a V-shaped bottom allow for maximum recovery of reagents. All reservoirs feature internal graduated volume lines and pour-spouts on the corners. Choose from opaque white polyvinyl chloride (PVC) or high clarity polystyrene (PS) in several packaging configurations.