Order Entry
ContactUsLinkComponent
Zeba Desalt Spin Columns 5 7K MWCO, 10mL
Pierce Zeba™ Desalt Spin Columns, Thermo Scientific
Catalog #: CAPI89891
Supplier:  Thermo Scientific
Zeba Desalt Spin Columns 5 7K MWCO, 10mL
Pierce Zeba™ Desalt Spin Columns, Thermo Scientific
Catalog #: CAPI89891
Supplier:  Thermo Scientific
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Capacity:
    5 ml
  • Description:
    Zeba™ Desalt Spin Columns, sufficient for 5 samples, each 500 - 2000 µL
  • Cat. no.:
    CAPI89891
  • Supplier No.:
    89891

Specifications

About this item

The Zeba™ spin desalting columns are made of low protein binding polypropylene and are compatible with a wide range of standard laboratory instruments and consumables.

  • Higher protein recovery
  • Minimal sample dilution
  • No screening fractions for protein or waiting for protein to emerge by gravity-flow
  • Easy-to-use with no cumbersome column preparation or equilibration

The Zeba 7K MWCO is recommended for processing proteins >7 kDa, and the Zeba 40K MWCO for processing proteins >40 kDa.

They provide >95% retention of salts and other small molecules.

Caution: Zeba™ columns are designed for research use only. Not for use in diagnostic procedures. Store at 4˚C.
Customers Who Bought This Also Bought