Order Entry
ContactUsLinkComponent
Hrv 3C Protease 1K-unit Kit
Human HRV 3C Protease Solution Kit, Pierce Chemical
Catalog #: CAPI88946
Supplier:  Thermo Scientific
CAS Number:  
Hrv 3C Protease 1K-unit Kit
Human HRV 3C Protease Solution Kit, Pierce Chemical
Catalog #: CAPI88946
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Species:
    Human
  • Size:
    1000 units
  • Storage conditions:
    Store at −20 °C
  • Enzyme name:
    HRV 3C protease solution kit
  • Enzyme activity:
    2 units/µl, 10000 units
  • Cat. no.:
    CAPI88946
  • Supplier No.:
    88946

Specifications

About this item

Thermo Scientific Pierce Human Rhinovirus (HRV) 3C Protease is a recombinant cysteine protease used to remove fusion tags from proteins with the HRV 3C cleavage sequence and is dual tagged for easy removal from the sample after cleavage.

  • Highly Specific with no Off-Target Cleavage Even at High Enzyme/Substrate Concentration
  • Allows Cleavage Reaction to Take Place at 4°C Without Affecting the Functionality of Purified Protein
  • Can be used Either Following Affinity Purification or While Fusion Proteins are Bound to Glutathione or a Tag-Specific Resin
  • Dual-Tagged Enzyme is Compatible with Various Columns and Buffers

Pierce HRV 3C Protease is highly specific for the recognition sequence Leu-Glu-Val-Leu-Phe-Gln-⊥-Gly-Pro and cleaves after the glutamine residue

Alternative names: Human Rhinovirus 3C
Source: Escherichia coli (E.coli)
Molecular weight: 47.8kD
Form: Liquid
Protease type: Cysteine
Optimal reaction conditions: 50 mM Tris-HCl, 150 mM NaCl, pH 8.0

The recombinant enzyme is engineered as a fusion protein with glutathione-S-transferase (GST) and polyhistidine (6xHis) tags, so that the protease can be easily removed after cleavage using a variety of solid supports with immobilized glutathione (GSH) or metal chelates, respectively. HRV3C is also active in a variety of commonly used buffers, providing further flexibility in experimental design.

Delivery: The kit contains HRV 3C protease (2 units/µl), reaction buffer and GST-Syk fusion protein.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought