Order Entry
ContactUsLinkComponent
M-PER Mammalian Protein Extraction Reagent 250mL
Pierce M-PER™, Mammalian Protein Extraction Reagents, Thermo Scientific
Catalog #: CAPI78501
Supplier:  Thermo Scientific
CAS Number:  
M-PER Mammalian Protein Extraction Reagent 250mL
Pierce M-PER™, Mammalian Protein Extraction Reagents, Thermo Scientific
Catalog #: CAPI78501
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    M-PER™ Mammalian Protein Extraction Reagents, for 25 g of Wet Cells
  • Size:
    250 mL
  • Cat. no.:
    CAPI78501
  • Application:
    Luciferase, b-Galactosidase (Low Signal), CAT, and Kinase Assays; ELISAs; Immobilized Glutathione; Western Blots
  • Supplier No.:
    78501

Specifications

About this item

M-PER™ Mammalian Protein Extraction Reagent is designed to provide highly efficient total protein extraction from cultured mammalian cells.

  • Gentle - mild detergent lysis, yielding extracts that are immediately compatible with Coomassie (Bradford) and BCA protein assays or SDS-PAGE
  • Compatible - extracts soluble proteins in non-denatured state, enabling direct use in immunoprecipitation and other affinity purification procedures
  • Amine-free and dialyzable - formulation ensures compatibility with subsequent assay systems
  • Convenient - lyse adherent cells directly in plate or after scraping and washing in suspension
  • Non-denaturing - maintain activity of luciferase, beta-galactosidase, CAT and other reporter genes as well or better than alternative methods

The complete cell lysis reagent dissolves cell membranes and extracts total cellular protein in only 5 minutes. This reagent requires little or no mechanical disruption and yields more protein than freeze/thaw cycles and sonication. This mammalian cell lysis reagent is so efficient that adherent cells do not need to be scraped from the culture dish, enabling easy and direct lysis and analysis of cells grown in 24-well and 96-well plates. Resulting cell lysates from either adherent and suspension cells are compatible with many downstream assays including immunoassays, enzyme assays and a variety of common reporter assays.

Customers Who Bought This Also Bought