Order Entry
ContactUsLinkComponent
Halt Protease and Phosphatase Inhibitor Cocktail, EDTA-free (100X) 10mL
Halt™ Protease and Phosphatase Inhibitor Cocktails, Thermo Scientific
Catalog #: CAPI78447
Supplier:  Thermo Scientific
CAS Number:  
Halt Protease and Phosphatase Inhibitor Cocktail, EDTA-free (100X) 10mL
Halt™ Protease and Phosphatase Inhibitor Cocktails, Thermo Scientific
Catalog #: CAPI78447
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Halt Protease and Phosphatase Inhibitor, Single-Use Cocktail, EDTA-free, (100 X)
  • Size:
    10 mL
  • Storage conditions:
    Upon receipt store at 4˚C. Product shipped with an ice pack.
  • Cat. no.:
    CAPI78447
  • Supplier No.:
    78447

Specifications

About this item

Halt Protease and Phosphatase Inhibitor Cocktail (100X) provides the convenience of a single solution with full protein sample protection for cell and tissue lysates.

  • 100X liquid formulation is easier and more effective to use than individual inhibitors
  • Aqueous-based format allows for convenient cold-room storage
  • Free of AEBSF and can be used in Mass spectrometry
  • Compatible with Thermo Scientific Pierce Cell Lysis Buffers

This Halt Protease and Phosphatase Inhibitor Cocktail is specifically optimized to protect proteins from degradation during extraction and purification. The cocktail contains inhibitors against the major classes of proteases and phosphatases, targeting aminopeptidases, cysteine and serine proteases and serine/threonine and protein tyrosine phosphatases.

Customers Who Bought This Also Bought