Pierce™ Bacterial Protein Extraction Reagents, Thermo Scientific
Catalog #: CAPI78243
Supplier: Thermo Scientific
CAS Number:
Specifications
- Description:B-PER® Bacterial Protein Extraction Reagent
- Size:165 mL
- Cat. no.:CAPI78243
- Supplier No.:78243
Specifications
About this item
Thermo Scientific B-PER Bacterial Protein Extraction Reagents are designed to extract soluble protein from bacterial cells without harsh chemicals or mechanical disruption.
- Lysis of bacteria occurs directly in cell culture media
- No centrifugation steps
- Compatible with the addition of protease inhibitors, and the resulting protein extract can be used in protein assays, typical affinity purification methods (eg. GST, 6XHis), and other applications
These easy-to-use cell lysis reagents are nonionic detergent solutions that effectively disrupt cells and solubilize native or recombinant proteins without denaturation.
Certain formulations include DNase I and lysozyme enzyme supplements to improve yield of large molecular weight proteins that are difficult to purify. B-PER Reagents are compatible with the addition of standard protease inhibitors and with downstream protein purification methods, such as GST purification and His-tagged immobilized metal affinity chromatography (IMAC).
Delivery: Kit contains B-PER® Reagent, 250 mL; Lysozyme, 50 mg/mL, 0.5 mL; and DNase I, 2500 U/mL, 0.5 mL. Separate reagents are also available.
Customers Who Bought This Also Bought