Specifications
- Description:PBS with Tween® 20
- Size:1 L
- Cat. no.:CAPI37539
- Application:ELISA, Western Blot, Dot Blot, Immunohistochemistry
- Supplier No.:37539
Specifications
About this item
These StartingBlock™ blocking buffers are PBS and TBS formulations of a proprietary protein that provides exceptional performance and broad compatibility with membrane (Western blot) and 96-well plate (ELISA) assays.
- Compatible with many detection systems (Western blot, ELISA and IHC)
- Short blocking times
- High signal:noise ratio
- Strip and reprobe without reblocking
- Available ready to use 1X formulations in PBS and TBS
StartingBlock Blocking Buffers performs well with an extremely wide range of antibodies, antibody combinations, other protein probing and assay systems.
These buffers are biotin- and serum-free. They are compatible with many detection systems and feature short blocking times. These buffers also allow stripping and reprobing with reblocking to save time and money.