Order Entry
ContactUsLinkComponent
SuperBlock* T20 (PBS) Blocking Buffer 1l
SuperBlock™ T20 (PBS) Blocking Buffer, Thermo Scientific
Catalog #: CAPI37516
Supplier:  Thermo Scientific
CAS Number:  
SuperBlock* T20 (PBS) Blocking Buffer 1l
SuperBlock™ T20 (PBS) Blocking Buffer, Thermo Scientific
Catalog #: CAPI37516
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    PBS with Tween® 20
  • Size:
    1 L
  • Cat. no.:
    CAPI37516
  • Application:
    ELISA, Western Blot, Dot Blot, Immunohistochemistry, Hybridization
  • Supplier No.:
    37516

Specifications

About this item

SuperBlock™ Blocking Buffer is optimized solution containing a single purified glycoprotein that provides incredibly fast and effective blocking for ELISA, IHC, and western blot analysis.

  • Blocking time—block membranes in 5 to 10 minutes and ELISA plates in 2 minutes
  • Applications—ELISA, western blotting, IHC; biotin-free for use with streptavidin system
  • Stability—store buffer at 4 °C for one year; store blocked plates dry for up to 12 months

Blocking micro-plates, membranes or tissues with a SuperBlock Buffer yields a high signal-to-noise ratio in most detection systems. The protein-based formulation does not contain any immunoglobulins, albumin, or endogenous biotin, making it compatible in many situations where traditional blocking agents fail. The buffer is especially effective at blocking coated polystyrene microplates (96-well plates) and stabilizing them for drying and storage for later use.

Customers Who Bought This Also Bought