Order Entry
ContactUsLinkComponent
1-Step Ultra TMB-ELISA Substrate Solution
Pierce™ 1-Step™ Ultra TMB-ELISA Substrate Solution, Thermo Scientific
Catalog #: CAPI34028
Supplier:  Thermo Scientific
CAS Number:  
1-Step Ultra TMB-ELISA Substrate Solution
Pierce™ 1-Step™ Ultra TMB-ELISA Substrate Solution, Thermo Scientific
Catalog #: CAPI34028
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugate ELISA:
    HRP
  • Sensitivity:
    VeryHigh
  • Description:
    1-Step™ Ultra TMB-ELISA
  • Size:
    250 mL
  • Cat. no.:
    CAPI34028
  • Supplier No.:
    34028

Specifications

About this item

The 1-Step™ Ultra TMB-ELISA contains soluble TMB (3,3´,5,5´-tetramentylbenzidine) that is ideal for sensitive ELISA-based applications

  • Available in traditional or ready-to-use formulations
  • No additional reagents or filtering required
  • Non carcinogenic
  • Upon receipt store at 4˚C, protected from moisture
  • Product is shipped at ambient temperature

TMB is a chromogenic substrate for horseradish peroxidase. Once oxidised by the enzyme, this substrate yields a blue product that absorbs at 370 nm and 652 nm. Adding sulfuric or phosphoric acid to stop the reaction changes the product to yellow that absorbs at 450 nm and provides greater sensitivity than the blue product. The 1-Step Ultra TMB-ELISA is supplied ready to use with no mixing required.

Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought