Order Entry
ContactUsLinkComponent
Reagent NHS-PEG4-Azide Ligation W/Spcr
Pierce™ NHS-Azide and NHS-Phosphine Reagents, Thermo Scientific
Catalog #: CAPI26130
Supplier:  Thermo Scientific
CAS Number:  
Reagent NHS-PEG4-Azide Ligation W/Spcr
Pierce™ NHS-Azide and NHS-Phosphine Reagents, Thermo Scientific
Catalog #: CAPI26130
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    NHS-PEG4-Azide
  • Size:
    100 mg
  • Storage conditions:
    Upon receipt store at –20 ˚C protected from light and moisture
  • Shipping temperature:
    ambient
  • Cat. no.:
    CAPI26130
  • Supplier No.:
    26130

Specifications

About this item

The NHS-Azide and NHS-Phosphine Reagents are amine-reactive compounds for derivatising primary amines of proteins or amine-coated polymer surfaces. Once a protein or surface is azide- or phosphine-labelled, the two components are mixed for effective and stable conjugation. Phosphine groups react with azides via a Staudinger reaction to produce an aza-ylide intermediate that is trapped to form a stable, covalent amide bond. Because phosphines and azides are absent from biological systems, there is minimal background labelling of macromolecules found in cells or lysates.

  • Reagent Easily Dissolves in Water-Miscible Solvents for Subsequent Dilution in Aqueous Reaction Mixtures with Cell Lysates and Other Biological Samples
  • Compatible Reaction Chemistry Occurs Effectively in Simple Buffer Conditions; Requires no Accessory Reagents Such as Copper or Reducing Agents
  • NHS Esters are Specific for Covalent Attachment to Primary Amines (Side Chain of Lysine or N-terminus of Polypeptides)
  • Chemoselective Azide and Phosphine Groups do not React or Interfere with Components of Biological Samples (Bioorthogonal) but Conjugate to One Another with High Efficiency

This NHS-ester compound reacts to form covalent bonds with primary amines (e.g., side chain of lysine residues or aminosilane-coated surfaces). The azide (N3) group reacts with phosphine-labeled molecules by a mechanism known as Staudinger chemistry, enabling efficient and specific conjugation of derivatized molecules in biological samples.

Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought