Order Entry
ContactUsLinkComponent
BCA Protein Assay Kit
Pierce™ BCA® Protein Assay Kits and Reagents, Thermo Scientific
Catalog #: CAPI23227
Supplier:  Thermo Scientific
CAS Number:  
Pierce™ BCA® Protein Assay Kits and Reagents, Thermo Scientific
Catalog #: CAPI23227
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    BCA Protein Assay Kit
  • Size:
    500 ml
  • Cat. no.:
    CAPI23227
  • No. of Assays/Kit:
    250 tube assays or 2500 microplate assays
  • Supplier No.:
    23227

Specifications

About this item

Pierce BCA™ (bicinchoninic acid) reagents provide accurate determination of protein concentration with most sample types encountered in protein research. The Pierce BCA™ Protein Assay can be used to assess yields in whole cell lysates and affinity-column fractions, as well as to monitor protein contamination in industrial applications. Compared to most dye-binding methods, the BCA™ Assay is affected much less by protein compositional differences, providing greater protein-to-protein uniformity.

  • Colorimetric, so can measure with a standard spectrophotometer or plate reader (562 nm)
  • Excellent uniformity, exhibiting less protein-to-protein variation than dye-binding methods
  • Unaffected by typical concentrations of most ionic and nonionic detergents
  • Easier and faster than the classic Lowry method
  • High linearity and sensitivity—linear working range for BSA 20 to 2000 µg/mL; detection down to 5 µg/m (with the enhanced protocol)

The assay kits can be used to assess yields in whole cell lysates and affinity-column fractions, as well as to monitor protein contamination in industrial applications

The BCA protein assay combines the well-known reduction of Cu2+ to Cu1+ by protein in an alkaline medium with the highly sensitive and selective colorimetric detection of the cuprous cation (Cu1+) by bicinchoninic acid. The first step is the chelation of copper with protein in an alkaline environment to form a light blue complex. In this reaction, known as the biuret reaction, peptides containing three or more amino acid residues form a colored chelate complex with cupric ions in an alkaline environment containing sodium potassium tartrate.

In the second step of the color development reaction, bicinchoninic acid (BCA) reacts with the reduced (cuprous) cation that was formed in step one. The intense purple-colored reaction product results from the chelation of two molecules of BCA with one cuprous ion. The BCA/copper complex is water-soluble and exhibits a strong linear absorbance at 562nm with increasing protein concentrations. The BCA reagent is approximately 100 times more sensitive (lower limit of detection) than the pale blue color of the first reaction.

The reaction that leads to BCA color formation is strongly influenced by four amino acid residues (cysteine or cystine, tyrosine, and tryptophan) in the amino acid sequence of the protein. However, unlike the Coomassie dye-binding methods, the universal peptide backbone also contributes to color formation, helping to minimize variability caused by protein compositional differences.

Delivery: Each Pierce BCA™ protein assay kit contains BCA Reagent A (500 mL or 2x500 mL), BCA Reagent B (25 mL), and Albumin Standard Ampoules (2 mg/mL, 10x1 mL).
Customers Who Bought This Also Bought