Order Entry
ContactUsLinkComponent
Coomassie (Bradford) Protein Assay Kit
Pierce™ Coomassie (Bradford) Protein Assay Kits, Thermo Scientific
Catalog #: CAPI23200
Supplier:  Thermo Scientific
CAS Number:  
Coomassie (Bradford) Protein Assay Kit
Pierce™ Coomassie (Bradford) Protein Assay Kits, Thermo Scientific
Catalog #: CAPI23200
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Coomassie (Bradford) Protein Assay Kit
  • Size:
    630 Tube Assays/3800 Microplate Assays
  • Detectable Concentration:
    100 - 1500 µg/ml
  • Supplier No.:
    23200

Specifications

About this item

The Pierce™ Coomassie (Bradford) Protein Assay Kit is a ready-to-use, stable formulation of the traditional Bradford assay reagent to measure (A595 nm) total protein concentration compared to a protein standard.

  • Stable, ready-to-use kit of the classical Bradford assay reagent
  • Colorimetric measure with a standard spectrophotometer or plate reader (595 nm)
  • Easy to use, only single reagent; no working reagent preparation required
  • Almost immediate color development; add, mix, and read results
  • Detects protein concentration in the range 1 to 1,500 µg/mL
  • Microplate and cuvette protocols provided with the instructions and adaptable to several target working ranges

When Coomassie dye binds protein in an acidic medium, an immediate shift in absorption maximum occurs from 465nm to 595nm with a concomitant colour change from brown to blue.

Delivery: Kit contains Pierce™ Coomassie Protein Assay Reagent (950 mL); Albumin Standard Ampoules (2 mg/mL,10x1 mL).
Customers Who Bought This Also Bought