Order Entry
ContactUsLinkComponent
High Sensitivity Streptavidin-HRP 0.5ml
Pierce™ High Sensitivity Streptavidin-HRP, Thermo Scientific
Catalog #: CAPI21130
Supplier:  Thermo Scientific
CAS Number:  
High Sensitivity Streptavidin-HRP 0.5ml
Pierce™ High Sensitivity Streptavidin-HRP, Thermo Scientific
Catalog #: CAPI21130
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    High Sensitivity Streptavidin-HRP
  • Size:
    0.5 mL
  • Storage conditions:
    Storage at 4˚C for at least one year.
  • Cat. no.:
    CAPI21130
  • Supplier No.:
    21130

Specifications

About this item

Thermo Scientific Pierce High Sensitivity Streptavidin-HRP is an exclusive peroxidase-conjugated streptavidin biotin-binding protein that provides signal amplification like poly-HRP and exceptional storage stability.

  • Detect low levels of target without background; obtain high signal-to-noise ratios
  • Less conjugate in Western blotting and ELISA applications and still obtain excellent results
  • Compatible with typical chemiluminescent, fluorescent and colorimetric peroxidase substrates
  • Ready-to-use stabilized liquid format means there is no waiting for thawing and no need to aliquot
  • It can be diluted 8-to 10-fold more than traditional HRP conjugates and still produce similar results
Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought