To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Ultimate Grade Fetal Bovine Serum (FBS)
- Size:50 ml
- Hemoglobin:≤25 mg/dl
- Endotoxin:≤5 EU/ml
- Storage temperature:Frozen
- Cat. no.:89510-198
- Supplier No.:1600-050
Specifications
About this item
Ultimate Grade FBS is the ultimate in product quality and viral safety, undergoing additional BVD virus testing using qPCR technology.
- 100% US Origin
- Additional virus testing through qPCR technology
- Enhanced traceability and regulatory compliance
- ISIA Certified Supply Chain for Traceability
Industry-leading collection and production techniques provide additional features that elevate product quality, enhance traceability, and improve regulatory compliance.
Avantor Seradigm is a supplier of animal sera that provides the cell culture community with access to the most reliable supply of high performance, exceptional quality Fetal Bovine Serum (FBS). Our approach to sourcing and manufacturing results in product performance and quality unlike any other.