Order Entry
ContactUsLinkComponent
Time-Date Stamp Alarm Freezer Thermometer
Single Probe Thermometers with Alarm, Time Date Stamp, Thermco
Catalog #: 10025-450
Supplier:  Thermco
Time-Date Stamp Alarm Freezer Thermometer
Single Probe Thermometers with Alarm, Time Date Stamp, Thermco
Catalog #: 10025-450
Supplier:  Thermco

Specifications

  • Description:
    Thermometer, Freezers, certified at –20 °C
  • Probe:
    One 5 ml bottle, Bio-Safe glycol/water
  • Range:
    –50...+70 °C (...58...+158 °F)
  • Supplier No.:
    ACC821FREV

Specifications

About this item

These large digital thermometers simultaneously show the current, minimum, and maximum temperatures and update continuously.

  • Triple display
  • Visual LED and audible beeping alarms
  • Bio-safe glycol/water probe medium
  • Improved resolution to 0.1º
  • Standard size vaccine bottles
  • Min.-max. temp with time and date stamp

The probe is housed in an unbreakable plastic bottle filled with a buffering media that maintains the thermometer reading when the instrument door is opened. The thermometer and the probe bottle have dual magnets to attach to the instrument. Thermometers also feature a flip-open stand, wall mounting key hole, and an AAA alkaline battery.

An audible and visual alarm will trigger when the temperatures goes above or below the set temperatures. The minimum/maximum feature monitors critical temperature for any period of time.

Certifications: Each thermometer is certified and traceable to NIST. Each thermometer is serialized.
Customers Who Bought This Also Bought