2-Ethoxyethanol stabilized for synthesis, Sigma-Aldrich®
Catalog #: CA8.00857.1000
Supplier: MilliporeSigma
CAS Number: 110-80-5
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle
- Size:1 L
- Cat. no.:CA8.00857.1000
- Supplier No.:8.00857.1000
Specifications
Specification Test Results
- Solubility:(20 °C) soluble
- Melting Point:-100 °C
- Molar Mass:90.12 g/mol
- Boiling Point:135 °C (1013 hPa)
- Vapor Pressure:5 hPa (20 °C)
- Flash Point:40 °C
- Refractive Index:1.408 (20 °C, 589 nm)
- Explosion Limit:1.8 - 15.7 %(V)
- Density:0.93 g/cm3 (20 °C)
- pH:(H2O, 20 °C) neutral
- Ignition Point:235 °C
- Saturation concentration:18 g/m3 (20 °C) Air
- Evaporation number:43
- Hazard Class:3
- Hazard Class Description:Flammable Liquid
Specification Test Results
About this item
Synonyms: Ethylene glycol monoethyl ether
:2-Ethoxyethanol
Boiling Pt:135 °C (1013 hPa)
Melting Pt:–70 °C
Density:0.932 g/cm³ (20 °C)
ADR:3,III
:Length:
:Width:
:Height:
:Ø
:Thickness:
:M-Clarity™ quality level = MQ200
Flash Pt:40 °C (closed cup)
:Boiling Pt:
:Boiling Pt: 135 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –70 °C
:Flash Pt:
:Flash Pt: 40 °C (closed cup)
:M.W. 90.12 g/mol
Merck Index:13,03786
Storage Temperature:Ambient
EINECS:203-804-1
UN:1171
REACH:01-2119560582-38
:3
:III
:40 °C
:GHS02
:GHS06
:GHS08
:MQ200
Customers Who Bought This Also Bought