To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Size:1 L
- Storage conditions:Room temperature
- Cat. no.:89500-596
- Supplier No.:J833-1L
Specifications
About this item
LB Broth, Liquid is used for the propagation and maintenance of E.coli. Widely used for the preparation of plasmid DNA and recombinant proteins.
- Ready to use liquid media
- Clear, yellow to brown liquid
1L of media contains 10 grams of sodium chloride, 10 grams of tryptone/ cesein peptone, 5 grams of yeast extract bacteriological.
No growth detected in Bioburden (Milliflex) (EQC) test. pH @25°C is 6.65 - 7.15. It has refractive index of 1.334 - 1.342. Specific Gravity at 20°C is 1.0116 - 1.0128. Abs.@260nm (1:100, Water) is 0.2263 - 0.2663.