Order Entry
ContactUsLinkComponent
Chart 6In 7-Day Pk60
Charts and Pens for 6" Dickson® Chart Recorders, Graphic Controls
Catalog #: CA11032-004
Supplier:  Graphic Controls
Chart 6In 7-Day Pk60
Charts and Pens for 6" Dickson® Chart Recorders, Graphic Controls
Catalog #: CA11032-004
Supplier:  Graphic Controls

Specifications

  • Temperature range:
    –50 to 50°C/°F
  • Description:
    Chart Number 651
  • Cat. no.:
    CA11032-004
  • Supplier No.:
    31502696
  • Recording Time:
    7-day

Specifications

About this item

Graphic Controls supplies a full line of charts and markers to fit all available Dickson® recorders

Graphic Controls charts and markers provide precise fit and function for all available Dickson® recorders. Our products deliver OEM performance at factory direct savings.

Versatile and durable, Dickson® recorders are used in a wide variety of applications and conditions measuring temperature, humidity, and pressure. Graphic Controls charts and markers perfectly complement these diverse applications and conditions to ensure your critical data is accurately recorded and preserved.

Ordering information: For information on additional charts from Graphic Controls, please contact your Avantor sales representative.
Customers Who Bought This Also Bought