Order Entry
ContactUsLinkComponent
Beaker Cover For 1.5 Gl Unit, Holds (2) 400 Or 600Ml Beakers
Components for Branson Ultrasonic Cleaners, Branson Ultrasonics
Catalog #: 89500-244
Beaker Cover For 1.5 Gl Unit, Holds (2) 400 Or 600Ml Beakers
Components for Branson Ultrasonic Cleaners, Branson Ultrasonics
Catalog #: 89500-244

Specifications

  • Description:
    Beaker Cover (2-400 or 600 mL) for 5.7 L Unit
  • Cat. no.:
    89500-244
  • Supplier No.:
    100-032-520

Specifications

About this item

Customers Who Bought This Also Bought