WHEATON® Extended Controlled Dropper Tips, DWK Life Sciences
Catalog #: 89500-080
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Cap Size:15 mm
- Color:Green
- Cat. no.:89500-080
- Supplier No.:W242426-A
Specifications
About this item
Extended Controlled Dropper Tips are made of LDPE (Low-Density Polyethylene)
Extended Controlled Dropper Tips snap into the inside of the bottle neck and are available in bulk. Dropper tips can be covered with Wheaton dropper tip screw caps.
They dispense one drop at a time. The average drop size is 44µL (±6µL).
Customers Who Bought This Also Bought