VWR® Flexible Safety Caps
Catalog #: 89499-996
Supplier: VWR International
Specifications
- Color:Green
- Description:VWR® Flexible Safety Caps
- Cat. no.:89499-996
Specifications
About this item
Safety caps provide a safe and economical way to cap and recap blood collection tubes and disposable glass culture tubes, regardless of manufacturer.
- Caps are color-coded for easy identification
- Liquid-tight seals
- Safe and economical
VWR® Flexible Safety Caps are available for a wide range of biomedical and industrial applications and fit six different sized tubes. VWR® Flexible Safety Caps come in a variety of colors for coding samples. They feature liquid-tight seals to ensure sample integrity and prevent spillage of serum samples. Made in the USA.
Customers Who Bought This Also Bought