Pierce™ Western Blotting Filter Paper, Thermo Scientific
Catalog #: CAPI88600
Supplier: Thermo Scientific
Specifications
- Dimensions:8 x 10.5 cm
- Cat. no.:CAPI88600
- Type:Sheet
- Supplier No.:88600
- No. of Sheets:100
Specifications
About this item
Use to efficiently transfer protein from an electrophoresis gel to a membrane in northern, Southern, and Western blotting applications.
- High-quality
- Easy to use
- Two thicknesses
- Tested and recommended for use with various protein methods
Filter paper for Western blotting applications is available in various formats including precut sizes that match standard gel sizes.
Customers Who Bought This Also Bought