ContactUsLinkComponent
Pierce C18 Tips 100 Ul 96 Tips
Pierce™ C18 Pipette Tips, Thermo Scientific
Catalog #: CAPI87784
Supplier:  Thermo Scientific
Pierce C18 Tips 100 Ul 96 Tips
Pierce™ C18 Pipette Tips, Thermo Scientific
Catalog #: CAPI87784
Supplier:  Thermo Scientific
Specifications
  • Volume:
    100 µL
  • Tip Color:
    Transparent
  • Packed:
    96/tray
  • Cat. no.:
    CAPI87784
  • Tip Style:
    C18
  • For Use With:
    Mass spectrometer, microcentrifuge
  • Supplier No.:
    87784
Specifications
About this item
Pierce™ C18 Tips are ready-to-use pipette-tip columns of C18 resin that enable fast and efficient capture, concentration, desalting and elution of peptides for MALDI mass spectrometry and other methods.
  • Monolithic C18 fast-flow tips have low resistance and improved flow characteristics compared to other commercially available tips
  • Eight packs and 96-tip trays are automation compatible
Improve protein analysis results with Pierce™ C18 Tips by removing urea, salts and other contaminants before MS analysis.
These C18 tips offer excellent flow properties with a high-efficiency monolithic C18 sorbent for fast wetting, loading, washing and eluting. The five-step procedure is simple and requires less than 5 minutes to process protein digests, strong cation exchange fractions and other protein and peptide samples for mass spectrometric (MS) analyses.
The tips are ideal for preparing samples for matrix-assisted laser desorption ionization (MALDI) or nanoelectrospray ionization techniques. The Pierce™ C18 Tips are available in convenient 10 and 100 µl tips with binding capacities of 8 and 80 µg, respectively.
Ordering information: Store at room temperature.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought