Order Entry
ContactUsLinkComponent
Tubing Snakeskint Dialysis 10K
Pierce SnakeSkin™ Pleated Dialysis Tubing, MWCO 3500 to 10000, Thermo Scientific
Catalog #: CAPI68100
Supplier:  Thermo Scientific
Tubing Snakeskint Dialysis 10K
Pierce SnakeSkin™ Pleated Dialysis Tubing, MWCO 3500 to 10000, Thermo Scientific
Catalog #: CAPI68100
Supplier:  Thermo Scientific
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Volume/Length:
    3.8 mL/cm
  • Cat. no.:
    CAPI68100
  • Inner Diameter:
    22 mm
  • MWCO:
    10 kDa
  • Supplier No.:
    68100

Specifications

About this item

Simplifies dialysis of sample volumes between 15 ml and 100 ml.

  • Ready to use
  • Pre-opened and pre-wetted
  • Greater than 90% protein recovery

Tubing is made of regenerated cellulose manufactured by the viscose process, which is then pleated/compressed into a hollow stick.

To use, simply pull out the required length, cut, and apply closure to either end.

Add the sample directly to the other end of the dry tubing, then apply another closure.

Pleated format makes tubing easy to open and use. Performance is similar to that of traditional dry, flat tubing.

Delivery: Supplied in eight-inch (20 cm) cylinders containing 35 feet (10.7 m) of tubing.
Customers Who Bought This Also Bought