ContactUsLinkComponent
Tape Sealing F/96Well Pk100
Sealing Tape for 96-Well Plates, Thermo Scientific
Catalog #: CAPI15036
Supplier:  Thermo Scientific
Tape Sealing F/96Well Pk100
Sealing Tape for 96-Well Plates, Thermo Scientific
Catalog #: CAPI15036
Supplier:  Thermo Scientific
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Sealing tape, clear acetate with acrylic adhesive, pre-cut sheets, for 96-well microplates
  • Cat. no.:
    CAPI15036
  • Supplier No.:
    15036
Specifications
About this item
Thermo Scientific Microplate Sealing Tape is a package of pre-cut, pressure-sensitive sheets for sealing the surface of standard 96-well or 384-well microplates during incubation steps.
  • Pressure-sensitive—position the tape over plate and press to seal; easy to remove
  • Separate sheets—each sheet of tape is supplied on its own easy-to-peal backing (liner)
  • Durable and inert
Use the disposable sheets to seal and reseal a microplate two or three times for incubation steps in an ELISA or protein assay procedure. Seal plates quickly and easily to allow mixing or prevent evaporation during assay incubation steps.
Caution: For Research Use Only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought