Order Entry
ContactUsLinkComponent
Silicone Tubing 2.5M 3Mm Id
Accessories for VACUSIP Bench-Top Aspiration System, INTEGRA Biosciences
Catalog #: 89498-878
Accessories for VACUSIP Bench-Top Aspiration System, INTEGRA Biosciences
Catalog #: 89498-878

Specifications

  • Description:
    Silicone Tubing, 2.5 m Length, 3 mm I.D.
  • Cat. no.:
    89498-878
  • Supplier No.:
    171023

Specifications

About this item

Customers Who Bought This Also Bought