To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Capacity:3 L
- Sterility:Non sterile
- Description:24-Hour Urine Container
- Packaging:Bulk Packed
- Supplier No.:30 3000
Specifications
About this item
Alleviate mess and exposure when handling urine collection and sampling with the Samco™ 24-Hour Urine Container range
- Range of sizes: 3000mL and 3500mL
- Amber tinted to protect from light exposure
- Easy to read, accurate graduations
- Custom cap and mouth design helps prevent leakage
- Large carrying handle for ease of transportation
Containers are amber tinted to protect from light exposure, and they are also inert to normal preservatives.