To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Capacity:90 ml
- Sterility:Non sterile
- Packaging:4 x Bag of 100
- Label Type:No label
- Cap Color:Yellow
- Supplier No.:02 0147
Specifications
About this item
Prevent bio-hazardous spills while conserving laboratory space with stackable Samco™ Bio-Tite™ Narrow-Mouth 90mL (3oz.) 48mm Specimen Containers
- Plug seal cap ensures excellent leak-resiatant performance
- Cap and base knurls ensure ease of handling
- Containers are stackable on top of each other for ease of storage
- Supplied nonsterile or gamma irradiated for guaranteed sterility
These shatter-resistant containers feature a leak-resistant seal and are ideal for the collection and transport of both liquid and solid samples. Available in both sterile and nonsterile formats, with a range of cap colors and label types to suit most applications.