To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Sterility:Sterile
- Description:Cap and Container Pre-Assembled (includes Sodium Thiosulfate Tablet)
- Cat. no.:89498-288
- Label Type:Non-tabbed
- Cap Color:White
- Supplier No.:01 0051
Specifications
About this item
Samco™ Wide-Mouth Bio-Tite™ Specimen Containers are prevent biohazardous spills with the proprietary, all plastic, leak-resistant seal.
All containers have a 120 ml (4oz.) capacity and a 53mm closure size. Wide-mouth containers are ideally suited for urine collection from women during pregnancy and for older or overweight patients. These stackable, shatter-resistant containers are manufactured from virgin polypropylene and feature the Bio-Tite™ system. They are tested at varying pressures, making them ideal for use in various transport systems.