Order Entry
ContactUsLinkComponent
Cap, Flange Plug, 12Mm, Black
Flanged Plug Caps, 12 mm, Globe Scientific
Catalog #: 89497-868
Supplier:  Globe Scientific
Cap, Flange Plug, 12Mm, Black
Flanged Plug Caps, 12 mm, Globe Scientific
Catalog #: 89497-868
Supplier:  Globe Scientific

Specifications

  • Color:
    Black
  • Description:
    Flanged Plug Cap
  • Material:
    Polyethylene
  • Cat. no.:
    89497-868
  • For Tube O.D.:
    12 mm
  • Supplier No.:
    118127K

Specifications

About this item

This line of polyethylene flanged plug caps provides exceptional leak-resistance in test tubes, centrifuge tubes, and round cuvettes.

  • Low density polyethylene (LDPE)
  • Assorted colors available
  • Fits both glass and plastic tubes

Flanged plug caps feature two flexible flanges that press firmly against the inside wall of the tubes, thereby providing better leak-resistance than standard plug caps. Available in a variety of colors to facilitate identification.

Customers Who Bought This Also Bought