Order Entry
ContactUsLinkComponent
Test Tube, 16 X 100Mm (12Ml), polyproylene With Rim, Graduated At 2.5, 5 & 10Ml
Polystyrene and Polypropylene Test Tubes, 16×100 mm, Globe Scientific
Catalog #: 89497-830
Supplier:  Globe Scientific
Test Tube, 16 X 100Mm (12Ml), polyproylene With Rim, Graduated At 2.5, 5 & 10Ml
Polystyrene and Polypropylene Test Tubes, 16×100 mm, Globe Scientific
Catalog #: 89497-830
Supplier:  Globe Scientific

Specifications

  • Diameter ×L:
    16 x 100 mm
  • Volume:
    12 ml
  • Description:
    Test Tube, with Rim, Graduated at 2.5mL, 5mL, and 10mL
  • Material:
    PP
  • Supplier No.:
    111040

Specifications

About this item

These tubes are molded from either premium-grade polystyrene for clarity, or polypropylene for impact strength and stability to common acids, alkalis, and solvents.

Customers Who Bought This Also Bought