Order Entry
ContactUsLinkComponent
Test Tube, 16 X 100Mm (12Ml), polystyrene No Rim, 500/Bag, 4 Bags/Unit
Polystyrene and Polypropylene Test Tubes, 16×100 mm, Globe Scientific
Catalog #: 89497-828
Supplier:  Globe Scientific
Test Tube, 16 X 100Mm (12Ml), polystyrene No Rim, 500/Bag, 4 Bags/Unit
Polystyrene and Polypropylene Test Tubes, 16×100 mm, Globe Scientific
Catalog #: 89497-828
Supplier:  Globe Scientific

Specifications

  • Diameter ×L:
    16 x 100 mm
  • Volume:
    12 ml
  • Description:
    Test Tube, without Rim
  • Material:
    Polystyrene
  • Supplier No.:
    111012

Specifications

About this item

These tubes are molded from either premium-grade polystyrene for clarity, or polypropylene for impact strength and stability to common acids, alkalis, and solvents.

Customers Who Bought This Also Bought