Polystyrene and Polypropylene Test Tubes, 17×100 mm, Globe Scientific
Catalog # 89497-784
Supplier: Globe Scientific
Specifications
- Ø×L:17×100 mm
- Volume:Sterile culture tube, with attached dual position cap
- Description:Sterile Culture Tube, with Attached Dual Position Cap
- Material:PP
- Cat. no.:89497-784
- Supplier No.:110178
Specifications
About this item
These tubes are molded either from premium-grade polystyrene for clarity, or polypropylene for impact strength and stability to common acids, alkalis and solvents.
Caps are made of polyethylene and are not autoclavable.
Frequently Bought Together