Specifications
- Capacity:5.0 ml
- Sterility:Non sterile
- Color:Colorless
- Environmentally Preferable:Low Manufacturing Impact
- Material:PP
- Environmental certification:Renewable Energy Certificate, ISO 14001
- Packaging:100 tubes (2 bags × 50 tubes)
- Style:PCR clean
- Maximum RCF:25000×g
- Supplier No.:0030108302
Specifications
About this item
Engineered to minimize sample binding and elevate recovery, Eppendorf® Protein LoBind® tubes safeguard valuable proteins during processing. The cutting-edge LoBind® technology ensures optimal yields for Western blotting, mass spectrometry, and protein purification applications.
- Surface-coating-free, including substances like silicone, to reduce the risk of sample interference
- Lot-certified PCR clean purity grade guarantees freedom from human DNA, DNase, RNase, and PCR inhibitors
- Available in tube, microplate, and deep-well plate formats for convenient upscaling of experiments
- Snap, screw, and SnapTec® cap options available
Unlock the benefits of Eppendorf® Protein LoBind® tubes, which are meticulously designed to offer unparalleled advantages. Available in a range of sizes and formats, these tubes are precision-crafted to minimize sample binding, providing a protective shield for valuable proteins, particularly in low-concentration applications. The LoBind® technology not only diminishes the risk of sample loss but also elevates protein recovery, guaranteeing users extract the maximum yield from every precious sample. Gain peace of mind knowing that proteins are safeguarded and results are optimized with Eppendorf® Protein LoBind® Tubes.
Applications include preparation or storage of protein, peptide, or antibody samples, Enzymatic assays (the hydrophilic surface reduces denaturation when it comes into contact with the tube inner wall), prevention of sample loss during storage of virus stock solutions, and storage of cell suspensions.
Manufactured without the use of slip agents, plasticizers, and biocides - substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.