Specifications
- Cap Type:Domed
- Capacity:0.2 ml
- Color:Natural
- Description:PCR reaction strips with attached cap
- Cat. no.:CA11028-446
- Supplier No.:T320-3N
Specifications
About this item
This 0.2 ml tube strip incorporates individually attached caps, eliminating the need to carry two separate components in inventory.
- Autoclavable
This PCR reaction strip is available with either flat or dome-topped individually attached hinged caps. While easily opened and closed with one hand, their positive sealing will fully protect the contents from evaporation during the entire thermal cycle. The cap has an integral seal preventing contamination with the lid surface.
Designed for easier handling than single tubes, the strip will precisely fit standard well spacing and can also be used with 8-channel hand-held pipettors. They are manufactured under the most stringent conditions to attain the highest quality industry standards. Available in a choice of colorless or four non-cytotoxic and non-metallic color varieties.
The strip includes 8 integral 0.2 ml tubes with ultra-thin sidewalls and bottoms for more uniform and efficient temperature transfer.