VWR® Sterile Bucket Liners
Catalog #: 89495-436
Supplier: VWR International
Small Business Enterprise
Specifications
- Description:Fits 1 gal. buckets, sterile
- Cat. no.:89495-436
Specifications
About this item
These LDPE/nylon blend bucket liners/bags create an impermeable barrier between strong disinfectant solutions and buckets.
- Sterile with sterility level of assurance (SAL 10⁻⁶)
- LDPE/nylon blend fabrication
- Provide an impermeable barrier against strong disinfectant solutions
This additional step ensures the integrity of the disinfectant blend. Item 89495-436 can be used for transport of sterile equipment.
Customers Who Bought This Also Bought