VWR® B.O.D. Bottles without Barcode, 300 ml
Catalog #: 89429-198
Supplier: VWR International
Specifications
- Capacity:300 ml (10.1 oz.)
- Number Sequence:Unnumbered
Specifications
About this item
Bottles conform to ASTM E438, Type I, Class B, borosilicate glass.
Customers Who Bought This Also Bought