Accessories for VWR® Filter Holder, 47 mm, 40/35 Joint Connection
Catalog #: 89428-982
Supplier: VWR International
Accessories for VWR® Filter Holder, 47 mm, 40/35 Joint Connection
Catalog #: 89428-982
Supplier: VWR International
Specifications
- Description:VWR® Flask Filter, 40/35, 1000 ml
- Cat. no.:89428-982
Specifications
About this item
Customers Who Bought This Also Bought
HYDRANAL® Karl Fischer Reagents, Honeywell Research Chemicals
Supplier: Honeywell Research Chemicals
$247.00 to $1,482.04
Sodium nitrate ≥99.0%, crystallised ACS, VWR Chemicals BDH®
Supplier: VWR International
$223.42 to $1,150.39
Buffers, Reference standards, Acidic to Neutral, Ricca Chemical
Supplier: Ricca Chemical
$200.31 to $2,030.49
Xylene (mixture of isomers), low water content 98.5-99.41% (isomers plus ethylbenzene) ACS, VWR Chemicals BDH®
Supplier: VWR International
$444.83 to $1,603.99
KIMBLE® Erlenmeyer Flasks, Jointed, Narrow Mouth, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
$105.21
Stannous chloride reagent II for phosphate analysis (with extraction)
Supplier: Ricca Chemical
$309.66