VWR® Nitrile 200 Examination Gloves
Catalog #: 89428-748
Supplier: VWR International
Specifications
- Ambidextrous/Hand-specific:Ambidextrous
- Glove Color:Blue
- Glove size:S
- Length:22.9 cm (9")
- Powdered/Powder-free:Powder-free
- Disposable / reusable:Disposable
- Finger thickness:3.1 mil
- Palm thickness:2.8 mil
- Cuff thickness:2.4 mil
- Cat. no.:89428-748
Specifications
About this item
These gloves are ambidextrous with textured fingers and a rolled, beaded cuff
- Ambidextrous, textured fingers, beaded cuff
- Tested and manufactured according to ISO 9001 and ISO 13485
- Thickness: 3.1 mil finger, 2.8 mil palm, and 2.4 mil cuff
Manufactured from 100% premium nitrile, these gloves provide hand protection against biohazards and a broad range of chemicals. These 100% latex-free gloves eliminate adverse reactions associated with natural rubber latex proteins, and have been tested for use with chemotherapy drugs per ASTM D6978. They are also rigorously tested and manufactured under stringent process controls according to ISO 9001 and ISO 13485 to ensure that each product meets exacting quality standards.
They are made from premium nitrile for high quality and consistency.
Packaging: Gloves are packaged 200 per box to save space and reduce waste.
Customers Who Bought This Also Bought