Order Entry
ContactUsLinkComponent
Sension+ 5050T Portable Combination Ph Electrode For General Purpose Applications
sensION+ pH Electrodes, Hach
Catalog #: CA11028-178
Supplier:  Hach
Sension+ 5050T Portable Combination Ph Electrode For General Purpose Applications
sensION+ pH Electrodes, Hach
Catalog #: CA11028-178
Supplier:  Hach
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Length:
    85 mm
  • Description:
    sensION+ Portable pH electrode + ATC, 5050T for PH1, general use, plastic body
  • Cat. no.:
    CA11028-178
  • pH Range:
    0 - 14
  • Junction:
    Ceramic diaphragm
  • Connector:
    MP5
  • Supplier No.:
    LZW5050T97002

Specifications

About this item

To meet your analysis needs, combined pH electrodes are available with a variety of lengths, diameters and reference systems.

Our sensION+ high performance pH electrodes have encapsulated reference element, Ag⁺ ion barrier (except 5010T) and are combination type including built in Pt 1000 temperature sensor.

pH glass electrodes from Radiometer Analytical are made with highly shock-resistant glass, making them extremely tough. All pH probes may be used with sensION+ meters.

Certifications: CE marked. WEEE compliant.
Customers Who Bought This Also Bought