ContactUsLinkComponent
Formaldehyde Test Kit
Formaldehyde Test Kit, Model FM-1, Hach
Catalog #: CA11027-670
Supplier:  Hach
CAS Number:  
Formaldehyde Test Kit
Formaldehyde Test Kit, Model FM-1, Hach
Catalog #: CA11027-670
Supplier:  Hach
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Chemical method:
    Thymol blue
  • Method:
    Drop Count Titration/ Thymolphthalein
  • Cat. no.:
    CA11027-670
  • Supplier No.:
    2183100
  • No. of Tests:
    100
Specifications
About this item
Formaldehyde solutions are used for maintenance of reverse osmosis systems and as disinfectants
  • Utilizes drop count titration - a simple, economical titration method
  • Kit contains all required reagents and apparatus in a rugged carrying case for analysis in the field
Hazardous Items. Items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.
The Model FM-1 measures higher concentrations by drop count titration. Contains 100 tests.
Ordering information: Four reagents, clippers, bottle, droppers, instruction sheet, and carrying case included.
Customers Who Bought This Also Bought